Assay Details
Gene Symbol: DHRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750297.3 | XP_002750343.1 | 2 - 3 | 502 | 115 | |
XM_009000423.2 | XP_008998671.1 | 2 - 3 | 507 | 115 | ||
XM_017974335.1 | XP_017829824.1 | 2 - 3 | 367 | 115 | ||
XM_017974336.1 | XP_017829825.1 | 2 - 3 | 355 | 115 | ||
XM_017974337.1 | XP_017829826.1 | 2 - 3 | 168 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
