Assay Details
| Gene Symbol: IDH3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763401.4 | XP_002763447.1 | 5 - 6 | 540 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDH3G| Gene Symbol: IDH3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763401.4 | XP_002763447.1 | 5 - 6 | 540 | 92 | |
Set Membership: |
|