Assay Details
| Gene Symbol: POLDIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997207.2 | XP_008995455.2 | 7 - 8 | 773 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLDIP2| Gene Symbol: POLDIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997207.2 | XP_008995455.2 | 7 - 8 | 773 | 70 | |
Set Membership: |
|