Assay Details
| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965315.1 | XP_017820804.1 | 2 - 3 | 322 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTP4A3| Gene Symbol: PTP4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965315.1 | XP_017820804.1 | 2 - 3 | 322 | 87 | |
Set Membership: |
|