Assay Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743171.4 | XP_002743217.1 | 5 - 6 | 556 | 77 | |
| XM_009000755.2 | XP_008999003.1 | 6 - 7 | 584 | 77 | ||
| XM_009000759.2 | XP_008999007.1 | 6 - 7 | 584 | 77 | ||
| XM_009000763.2 | XP_008999011.1 | 5 - 6 | 556 | 77 | ||
| XM_009000776.2 | XP_008999024.1 | 6 - 7 | 585 | 77 | ||
| XM_017975225.1 | XP_017830714.1 | 6 - 7 | 583 | 77 | ||
| XM_017975233.1 | XP_017830722.1 | 5 - 6 | 453 | 77 | ||
More Information
Set Membership: |
|

FSD1L
Genomic Map