Assay Details
| Gene Symbol: DNASE1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763418.3 | XP_002763464.1 | 4 - 5 | 564 | 82 | |
| XM_008990099.2 | XP_008988347.1 | 4 - 5 | 569 | 82 | ||
| XM_008990100.2 | XP_008988348.1 | 5 - 6 | 941 | 82 | ||
More Information
Set Membership: |
|

DNASE1L1
Genomic Map