Assay Details
| Gene Symbol: CENPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746934.4 | XP_002746980.1 | 2 - 3 | 399 | 100 | |
| XM_017971376.1 | XP_017826865.1 | 2 - 3 | 663 | 100 | ||
More Information
Set Membership: |
|

CENPW
Genomic Map