Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983745.2 | XP_008981993.1 | 5 - 6 | 633 | 74 | |
| XM_017965792.1 | XP_017821281.1 | 5 - 6 | 10847 | 74 | ||
More Information
Set Membership: |
|

PXYLP1
Genomic Map