Assay Details
Gene Symbol: CCNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745682.3 | XP_002745728.1 | 3 - 4 | 364 | 90 | |
XM_008993124.2 | XP_008991372.1 | 3 - 4 | 927 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
