Assay Details
| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982262.2 | XP_008980510.1 | 7 - 8 | 1671 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PODXL2| Gene Symbol: PODXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982262.2 | XP_008980510.1 | 7 - 8 | 1671 | 64 | |
Set Membership: |
|