Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750072.3 | XP_002750118.1 | 13 - 14 | 1602 | 68 | |
| XM_008999997.2 | XP_008998245.1 | 14 - 15 | 1706 | 68 | ||
| XM_017974085.1 | XP_017829574.1 | 11 - 12 | 1430 | 68 | ||
| XM_017974086.1 | XP_017829575.1 | 9 - 10 | 1010 | 68 | ||
More Information
Set Membership: |
|

STAM
Genomic Map