Assay Details
Gene Symbol: C15H3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758341.2 | XP_002758387.1 | 2 - 3 | 741 | 75 | |
XM_017964903.1 | XP_017820392.1 | 3 - 4 | 647 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
