Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756750.3 | XP_002756796.1 | 3 - 4 | 1498 | 78 | |
| XM_008979005.2 | XP_008977253.1 | 3 - 4 | 1497 | 78 | ||
More Information
Set Membership: |
|

ANGPT2
Genomic Map