Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745018.4 | XP_002745064.1 | 5 - 6 | 3953 | 78 | |
| XM_008992114.2 | XP_008990362.1 | 5 - 6 | 825 | 78 | ||
| XM_008992116.2 | XP_008990364.1 | 5 - 6 | 973 | 78 | ||
| XM_008992117.2 | XP_008990365.1 | 5 - 6 | 3528 | 78 | ||
| XM_008992119.1 | XP_008990367.1 | 5 - 6 | 518 | 78 | ||
| XM_008992121.2 | XP_008990369.1 | 3 - 4 | 483 | 78 | ||
| XM_017969936.1 | XP_017825425.1 | 4 - 5 | 5211 | 78 | ||
More Information
Set Membership: |
|

GHR
Genomic Map