Assay Details
| Gene Symbol: CLEC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996632.2 | XP_008994880.1 | 8 - 9 | 1001 | 73 | |
| XM_008996633.2 | XP_008994881.1 | 8 - 9 | 1086 | 73 | ||
| XM_008996634.2 | XP_008994882.1 | 7 - 8 | 880 | 73 | ||
More Information
Set Membership: |
|

CLEC10A
Genomic Map