Assay Details
| Gene Symbol: CETP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760996.4 | XP_002761042.1 | 8 - 9 | 876 | 73 | |
| XM_008985919.2 | XP_008984167.1 | 8 - 9 | 803 | 73 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CETP| Gene Symbol: CETP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760996.4 | XP_002761042.1 | 8 - 9 | 876 | 73 | |
| XM_008985919.2 | XP_008984167.1 | 8 - 9 | 803 | 73 | ||