Assay Details
Gene Symbol: C1H22orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743539.3 | XP_002743585.1 | 1 - 2 | 2441 | 106 | |
XM_009006678.2 | XP_009004926.1 | 1 - 2 | 886 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002743539.3, XM_009006678.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
