Assay Details
| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744102.4 | XP_002744148.1 | 5 - 6 | 777 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THG1L| Gene Symbol: THG1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744102.4 | XP_002744148.1 | 5 - 6 | 777 | 80 | |
Set Membership: |
|