Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750052.4 | XP_002750098.2 | 4 - 5 | 1130 | 98 | |
| XM_008999969.2 | XP_008998217.1 | 4 - 5 | 1288 | 98 | ||
| XM_008999970.2 | XP_008998218.1 | 3 - 4 | 963 | 98 | ||
| XM_008999971.2 | XP_008998219.1 | 3 - 4 | 1008 | 98 | ||
| XM_008999972.2 | XP_008998220.1 | 3 - 4 | 1141 | 98 | ||
| XM_017974072.1 | XP_017829561.1 | 4 - 5 | 1130 | 98 | ||
More Information
Set Membership: |
|

SUV39H2
Genomic Map