Assay Details
| Gene Symbol: CKMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744843.4 | XP_002744889.1 | 7 - 8 | 953 | 74 | |
| XM_008991882.2 | XP_008990130.1 | 7 - 8 | 1006 | 74 | ||
More Information
Set Membership: |
|

CKMT2
Genomic Map