Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967533.1 | XP_017823022.1 | 5 - 6 | 1090 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERS1| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017967533.1 | XP_017823022.1 | 5 - 6 | 1090 | 85 | |
Set Membership: |
|