Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751026.2 | XP_002751072.1 | 3 - 4 | 636 | 61 | |
| XM_009001614.2 | XP_008999862.1 | 3 - 4 | 304 | 61 | ||
| XM_017974887.1 | XP_017830376.1 | 4 - 5 | 461 | 61 | ||
More Information
Set Membership: |
|

DDAH1
Genomic Map