Assay Details
| Gene Symbol: SMIM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757007.4 | XP_002757053.1 | 2 - 3 | 276 | 79 | |
| XM_008979403.2 | XP_008977651.1 | 2 - 3 | 240 | 79 | ||
| XM_008979404.2 | XP_008977652.1 | 2 - 3 | 218 | 79 | ||
| XM_008979405.2 | XP_008977653.1 | 2 - 3 | 253 | 79 | ||
| XM_008979406.2 | XP_008977654.1 | 2 - 3 | 184 | 79 | ||
| XM_008979407.2 | XP_008977655.1 | 2 - 3 | 184 | 79 | ||
| XM_017963825.1 | XP_017819314.1 | 2 - 3 | 207 | 79 | ||
More Information
Set Membership: |
|

SMIM19
Genomic Map