Assay Details
| Gene Symbol: PLAUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762212.3 | XP_002762258.1 | 2 - 3 | 392 | 88 | |
| XM_008988139.2 | XP_008986387.1 | 2 - 3 | 193 | 88 | ||
| XM_017968099.1 | XP_017823588.1 | 2 - 3 | 589 | 88 | ||
| XM_017968100.1 | XP_017823589.1 | 2 - 3 | 392 | 88 | ||
More Information
Set Membership: |
|

PLAUR
Genomic Map