Assay Details
| Gene Symbol: LOC100400332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998754.1 | XP_008997002.1 | 4 - 5 | 752 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100400332| Gene Symbol: LOC100400332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008998754.1 | XP_008997002.1 | 4 - 5 | 752 | 55 | |
Set Membership: |
|