Assay Details
Gene Symbol: CARD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006701.2 | XP_009004949.1 | 2 - 3 | 411 | 84 | |
XM_017977509.1 | XP_017832998.1 | 2 - 3 | 328 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
