Assay Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992455.2 | XP_008990703.1 | 3 - 4 | 342 | 81 | |
| XM_008992457.1 | XP_008990705.1 | 2 - 3 | 546 | 81 | ||
| XM_008992458.1 | XP_008990706.1 | 2 - 3 | 546 | 81 | ||
| XM_008992459.2 | XP_008990707.1 | 3 - 4 | 342 | 81 | ||
| XM_017970145.1 | XP_017825634.1 | 3 - 4 | 299 | 81 | ||
More Information
Set Membership: |
|

AADAT
Genomic Map