Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017969117.1 | XP_017824606.1 | 8 - 9 | 1037 | 64 | |
| XM_017969120.1 | XP_017824609.1 | 9 - 10 | 1089 | 64 | ||
More Information
Set Membership: |
|

FAXDC2
Genomic Map