Assay Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996570.2 | XP_008994818.1 | 2 - 3 | 307 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAPPC1| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996570.2 | XP_008994818.1 | 2 - 3 | 307 | 63 | |
Set Membership: |
|