Assay Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752930.3 | XP_002752976.1 | 2 - 3 | 698 | 58 | |
| XM_009004520.1 | XP_009002768.1 | 2 - 3 | 683 | 58 | ||
| XM_017976331.1 | XP_017831820.1 | 2 - 3 | 482 | 58 | ||
More Information
Set Membership: |
|

CHST11
Genomic Map