Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976662.1 | XP_017832151.1 | 4 - 5 | 1145 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PARP16| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976662.1 | XP_017832151.1 | 4 - 5 | 1145 | 59 | |
Set Membership: |
|