Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001335.1 | XP_008999583.1 | 6 - 7 | 1244 | 74 | |
| XM_009001336.2 | XP_008999584.1 | 6 - 7 | 1242 | 74 | ||
| XM_017974793.1 | XP_017830282.1 | 6 - 7 | 1244 | 74 | ||
More Information
Set Membership: |
|

OMA1
Genomic Map