Assay Details
| Gene Symbol: NIF3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749619.4 | XP_002749665.1 | 2 - 3 | 699 | 69 | |
| XM_008999177.2 | XP_008997425.1 | 2 - 3 | 554 | 69 | ||
| XM_008999178.2 | XP_008997426.1 | 2 - 3 | 699 | 69 | ||
More Information
Set Membership: |
|

NIF3L1
Genomic Map