Assay Details
| Gene Symbol: TNFRSF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733711.2 | XP_003733759.1 | 5 - 6 | 861 | 118 | |
| XM_017975877.1 | XP_017831366.1 | 5 - 6 | 568 | 118 | ||
More Information
Set Membership: |
|

TNFRSF1A
Genomic Map