Assay Details
| Gene Symbol: ADORA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007848.2 | XP_009006096.1 | 2 - 3 | 783 | 69 | |
| XM_009007852.2 | XP_009006100.1 | 2 - 3 | 709 | 69 | ||
More Information
Set Membership: |
|

ADORA2A
Genomic Map