Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759734.4 | XP_002759780.1 | 1 - 2 | 1748 | 101 | |
| XM_008984038.2 | XP_008982286.2 | 1 - 2 | 1748 | 101 | ||
More Information
Set Membership: |
|

MYD88
Genomic Map