Assay Details
| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745924.3 | XP_002745970.1 | 4 - 5 | 774 | 90 | |
| XM_008993495.2 | XP_008991743.1 | 4 - 5 | 800 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NWD2| Gene Symbol: NWD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745924.3 | XP_002745970.1 | 4 - 5 | 774 | 90 | |
| XM_008993495.2 | XP_008991743.1 | 4 - 5 | 800 | 90 | ||