Assay Details
Gene Symbol: SHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984481.2 | XP_008982729.2 | 4 - 5 | 1205 | 55 | |
XM_017966308.1 | XP_017821797.1 | 4 - 5 | 1205 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
