Assay Details
| Gene Symbol: REPIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751891.4 | XP_002751937.2 | 2 - 3 | 930 | 68 | |
| XM_009002844.2 | XP_009001092.1 | 2 - 3 | 365 | 68 | ||
More Information
Set Membership: |
|

REPIN1
Genomic Map