Assay Details
| Gene Symbol: LOC100392727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994994.2 | XP_008993242.2 | 69 - 70 | 9374 | 125 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100392727| Gene Symbol: LOC100392727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994994.2 | XP_008993242.2 | 69 - 70 | 9374 | 125 | |
Set Membership: |
|