Assay Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759828.4 | XP_002759874.2 | 6 - 7 | 855 | 86 | |
| XM_008984212.2 | XP_008982460.1 | 6 - 7 | 856 | 86 | ||
| XM_017966205.1 | XP_017821694.1 | 5 - 6 | 722 | 86 | ||
More Information
Set Membership: |
|

PRKAB2
Genomic Map