Assay Details
| Gene Symbol: STX16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747713.4 | XP_002747759.1 | 5 - 6 | 1473 | 60 | |
| XM_002747714.3 | XP_002747760.1 | 5 - 6 | 1524 | 60 | ||
| XM_002747715.3 | XP_002747761.1 | 4 - 5 | 1461 | 60 | ||
| XM_008996173.2 | XP_008994421.1 | 5 - 6 | 1524 | 60 | ||
| XM_008996174.2 | XP_008994422.1 | 4 - 5 | 1512 | 60 | ||
| XM_008996175.2 | XP_008994423.1 | 5 - 6 | 1473 | 60 | ||
| XM_008996176.2 | XP_008994424.1 | 4 - 5 | 1512 | 60 | ||
| XM_008996177.2 | XP_008994425.1 | 4 - 5 | 1461 | 60 | ||
More Information
Set Membership: |
|

STX16
Genomic Map