Assay Details
| Gene Symbol: GPER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744447.3 | XP_002744493.1 | 2 | 3728 | 97 | |
| XM_008989093.2 | XP_008987341.1 | 2 | 4177 | 97 | ||
More Information
Set Membership: |
|

GPER1
Genomic Map