Assay Details
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733929.3 | XP_003733977.1 | 6 - 7 | 737 | 65 | |
XM_009005473.2 | XP_009003721.1 | 5 - 6 | 618 | 65 | ||
XM_009005475.2 | XP_009003723.1 | 5 - 6 | 2785 | 65 | ||
XM_009005476.2 | XP_009003724.1 | 5 - 6 | 951 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
