Assay Details
| Gene Symbol: SEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009434.1 | XP_009007682.1 | 11 - 12 | 1197 | 74 | |
| XM_009009435.2 | XP_009007683.1 | 11 - 12 | 1164 | 74 | ||
More Information
Set Membership: |
|

SEPT1
Genomic Map