Assay Details
| Gene Symbol: CDH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985989.2 | XP_008984237.1 | 5 - 6 | 1012 | 64 | |
| XM_008985990.2 | XP_008984238.1 | 5 - 6 | 2021 | 64 | ||
More Information
Set Membership: |
|

CDH5
Genomic Map