Assay Details
| Gene Symbol: ACVRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752484.3 | XP_002752530.1 | 6 - 7 | 1101 | 69 | |
| XM_009003829.2 | XP_009002077.1 | 6 - 7 | 1106 | 69 | ||
More Information
Set Membership: |
|

ACVRL1
Genomic Map