Assay Details
| Gene Symbol: CHST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756258.3 | XP_002756304.2 | 2 - 3 | 1911 | 95 | |
| XM_009009763.2 | XP_009008011.1 | 2 - 3 | 581 | 95 | ||
More Information
Set Membership: |
|

CHST3
Genomic Map