Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733386.3 | XP_003733434.1 | 5 - 6 | 577 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM222| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733386.3 | XP_003733434.1 | 5 - 6 | 577 | 58 | |
Set Membership: |
|