Assay Details
| Gene Symbol: RSAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997373.2 | XP_008995621.1 | 1 - 2 | 1108 | 53 | |
| XM_008997375.2 | XP_008995623.1 | 1 - 2 | 1108 | 53 | ||
| XM_008997376.2 | XP_008995624.1 | 1 - 2 | 1108 | 53 | ||
More Information
Set Membership: |
|

RSAD1
Genomic Map